missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl 4-(6-Amino-3-pyridyl)piperazine-1-carboxylate 98.0+%, TCI America™

Catalog No. p-7123681
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B4669200MG 200 mg
B46691G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B4669200MG Supplier TCI America Supplier No. B4669200MG

Chemical Identifiers

CAS 571188-59-5
Molecular Formula C14H22N4O2
Molecular Weight (g/mol) 278.36
MDL Number MFCD11594962
InChI Key RMULRXHUNOVPEI-UHFFFAOYSA-N
Synonym tert-butyl 4-6-aminopyridin-3-yl piperazine-1-carboxylate, 1-boc-4-6-aminopyridin-3-yl piperazine, tert-butyl 4-6-amino-3-pyridyl piperazine-1-carboxylate, 1-boc-4-6-amino-3-pyridyl piperazine, 4-6-amino-3-pyridyl-1-boc-piperazine, 1-piperazinecarboxylic acid, 4-6-amino-3-pyridinyl-, 1,1-dimethylethyl ester, 1-tert-butoxycarbonyl-4-6-amino-3-pyridyl piperazine, t-butyl 4-6-aminopyridin-3-yl piperazine-1-carboxylate, tert-butyl4-6-aminopyridin-3-yl piperazine-1-carboxylate, 4-6-amino-3-pyridyl piperazine-1-carboxylic acid tert-butyl ester
PubChem CID 11737525
IUPAC Name tert-butyl 4-(6-aminopyridin-3-yl)piperazine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCN(CC1)C1=CN=C(N)C=C1

Specifications

Melting Point 133°C
Color Yellow
Quantity 200 mg
Formula Weight 278.36
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material tert-Butyl 4-(6-Amino-3-pyridyl)piperazine-1-carboxylate