missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl 4-[5-(hydroxymethyl)-4-methyl-1,3-thiazol-2-yl]piperidine-1-carboxylate, ≥97%, Thermo Scientific™

Catalog No. CC31109CB
Change view
Click to view available options
Quantity:
250mg
1g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC31109CB 250mg Amber Glass Bottle
CC31109DA 1g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC31109CB Supplier Thermo Scientific Chemicals Supplier No. CC31109CB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 857283-66-0
Molecular Formula C15H24N2O3S
Molecular Weight (g/mol) 312.428
MDL Number MFCD07368658
InChI Key YNLQKGMVENCCLE-UHFFFAOYSA-N
Synonym tert-butyl 4-5-hydroxymethyl-4-methyl-1,3-thiazol-2-yl tetrahydro-1 2h-pyridinecarboxylate, tert-butyl 4-5-hydroxymethyl-4-methyl-1,3-thiazol-2-yl piperidine-1-carboxylate, 1-piperidinecarboxylicacid, 4-5-hydroxymethyl-4-methyl-2-thiazolyl-, 1,1-dimethylethyl ester, 1-tert-butoxycarbonyl-4-5-hydroxymethyl-4-methyl-1,3-thiazol-2-yl piperidine, 4-5-hydroxymethyl-4-methyl-thiazol-2-yl-piperidine-1-carboxylic acid tert-butyl ester, 4-5-hydroxymethyl-4-methyl-1,3-thiazol-2-yl piperidine, n1-boc protected, tert-butyl 4-5-hydroxymethyl-4-methylthiazol-2-yl piperidine-1-carboxylate
PubChem CID 7164575
IUPAC Name tert-butyl 4-[5-(hydroxymethyl)-4-methyl-1,3-thiazol-2-yl]piperidine-1-carboxylate
SMILES CC1=C(SC(=N1)C2CCN(CC2)C(=O)OC(C)(C)C)CO

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 250mg
Formula Weight 312.43
Percent Purity 95%
Chemical Name or Material Tert-butyl 4-[5-(hydroxymethyl)-4-methyl-1, 3-thiazol-2-yl]tetrahydro-1(2H)-pyridinecarboxylate
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.