missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl 4-{4-[(methylamino)methyl]pyrid-2-yl}piperazine-1-carboxylate, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC50646DE

Quantity 5g
Packaging Amber Glass Bottle

 View more versions of this product

Catalog No. CC50646DE

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Chemical Identifiers

946409-15-0
306.41
BNLNDKXJNWJYKP-UHFFFAOYSA-N
28765107
CNCC1=CC(=NC=C1)N1CCN(CC1)C(=O)OC(C)(C)C
C16H26N4O2
MFCD09966148
tert-butyl 4-4-methylamino methyl pyridin-2-yl piperazine-1-carboxylate, tert-butyl 4-4-methylamino methyl pyrid-2-yl piperazine-1-carboxylate
tert-butyl 4-[4-(methylaminomethyl)pyridin-2-yl]piperazine-1-carboxylate

Specifications

tert-Butyl 4-{4-[(methylamino)methyl]pyrid-2-yl}piperazine-1-carboxylate
96.5
97%
C16H26N4O2
MFCD09966148
BNLNDKXJNWJYKP-UHFFFAOYSA-N
tert-butyl 4-[4-(methylaminomethyl)pyridin-2-yl]piperazine-1-carboxylate
28765107
97%
946409-15-0
100.0
Amber Glass Bottle
5g
tert-butyl 4-4-methylamino methyl pyridin-2-yl piperazine-1-carboxylate, tert-butyl 4-4-methylamino methyl pyrid-2-yl piperazine-1-carboxylate
CNCC1=CC(=NC=C1)N1CCN(CC1)C(=O)OC(C)(C)C
306.41
306.41
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS
Documents

Documents