missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl 4-{4-[(methylamino)methyl]pyrid-2-yl}piperazine-1-carboxylate, 97%, Thermo Scientific™

Catalog No. CC50646DA
Change view
Click to view available options
Quantity:
1g
5g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC50646DA 1g Amber Glass Bottle
CC50646DE 5g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC50646DA Supplier Thermo Scientific Chemicals Supplier No. CC50646DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 946409-15-0
Molecular Formula C16H26N4O2
Molecular Weight (g/mol) 306.41
MDL Number MFCD09966148
InChI Key BNLNDKXJNWJYKP-UHFFFAOYSA-N
Synonym tert-butyl 4-4-methylamino methyl pyridin-2-yl piperazine-1-carboxylate, tert-butyl 4-4-methylamino methyl pyrid-2-yl piperazine-1-carboxylate
PubChem CID 28765107
IUPAC Name tert-butyl 4-[4-(methylaminomethyl)pyridin-2-yl]piperazine-1-carboxylate
SMILES CNCC1=CC(=NC=C1)N1CCN(CC1)C(=O)OC(C)(C)C

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1g
Formula Weight 306.41
Percent Purity 97%
Chemical Name or Material tert-Butyl 4-{4-[(methylamino)methyl]pyrid-2-yl}piperazine-1-carboxylate
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Harmful if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.