missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl 4-(4-formyl-1,3-thiazol-2-yl)piperidine-1-carboxylate, ≥97%, Thermo Scientific™

Catalog No. CC31004CB
Change view
Click to view available options
Quantity:
250mg
1g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC31004CB 250mg Amber Glass Bottle
CC31004DA 1g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC31004CB Supplier Thermo Scientific Chemicals Supplier No. CC31004CB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 869901-02-0
Molecular Formula C14H20N2O3S
Molecular Weight (g/mol) 296.385
MDL Number MFCD06658981
InChI Key XLGKMJFDRZHAEV-UHFFFAOYSA-N
Synonym tert-butyl 4-4-formylthiazol-2-yl piperidine-1-carboxylate, tert-butyl 4-4-formyl-1,3-thiazol-2-yl piperidine-1-carboxylate, tert-butyl 4-4-formyl-1,3-thiazol-2-yl tetrahydro-1 2h-pyridinecarboxylate, tert-butyl 4-4-formylthiazol-2-yl piperidine-1-ca, tert-butyl4-4-formylthiazol-2-yl piperidine-1-carboxylate, 1,1-dimethylethyl 4-4-formyl-2-thiazolyl-1-piperidinecarboxylate, 4-4-formyl-thiazol-2-yl-piperidine-1-carboxylic acid tert-butyl ester, 1-piperidinecarboxylicacid, 4-4-formyl-2-thiazolyl-, 1,1-dimethylethyl ester, 4-4-formyl-2-thiazolyl-1-piperidinecarboxylic acid,1,1-dimethylethyl ester
PubChem CID 7172298
IUPAC Name tert-butyl 4-(4-formyl-1,3-thiazol-2-yl)piperidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCC(CC1)C2=NC(=CS2)C=O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250mg
Formula Weight 296.39
Percent Purity 97%
Chemical Name or Material tert-Butyl 4-(4-formyl-1, 3-thiazol-2-yl)tetrahydro-1(2H)-pyridinecarboxylate
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.