missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl 4-(4-Bromopyrazol-1-yl)piperidine-1-carboxylate 98.0+%, TCI America™

Catalog No. B43915G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B43915G 5 g
B43911G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B43915G Supplier TCI America Supplier No. B43915G

Chemical Identifiers

CAS 877399-50-3
Molecular Formula C13H20BrN3O2
Molecular Weight (g/mol) 330.23
MDL Number MFCD09831980
InChI Key IYNZAVDBHAQODX-UHFFFAOYSA-N
Synonym tert-butyl 4-4-bromo-1h-pyrazol-1-yl piperidine-1-carboxylate, 1-boc-4-4-bromopyrazol-1-yl piperidine, 4-4-bromopyrazol-1-yl piperidine-1-carboxylic acid tert-butyl ester, 4-4-bromo-pyrazol-1-yl-piperidine-1-carboxylic acid tert-butyl ester, tert-butyl 4-4-bromopyrazol-1-yl piperidine-1-carboxylate, n-boc-4-4-bromopyrazol-1-yl piperidine, 3-2-ethoxycarbonylacetyl piperidine-1-carboxylic acid tert-butyl ester, 1-piperidinecarboxylicacid, 4-4-bromo-1h-pyrazol-1-yl-, 1,1-dimethylethyl ester, ksc658s5d, 1-boc-4-4-bromo-1-pyrazolyl piperidine
PubChem CID 45480278
IUPAC Name tert-butyl 4-(4-bromo-1H-pyrazol-1-yl)piperidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCC(CC1)N1C=C(Br)C=N1

Specifications

Melting Point 79°C
Color White
Quantity 5 g
Formula Weight 330.23
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material tert-Butyl 4-(4-Bromopyrazol-1-yl)piperidine-1-carboxylate