missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl 4-[4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)pyrazol-1-yl]piperidine-1-carboxylate 98.0+%, TCI America™

Catalog No. p-7123893
Change view
Click to view available options
Quantity:
1 g
5 g
Catalog No. Quantity
B50021G 1 g
B50025G 5 g
2 options

Catalog No. B50021G

Supplier: TCI America B50021G

Only null left

Chemical Identifiers

CAS 877399-74-1
Molecular Formula C19H32BN3O4
Molecular Weight (g/mol) 377.292
MDL Number MFCD11112131
InChI Key QSQWENQPOSRWLP-UHFFFAOYSA-N
Synonym 4-[4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)pyrazol-1-yl]piperidine-1-carboxylic Acid tert-Butyl Ester, 2-[1-(1-Boc-piperidin-4-yl)pyrazol-4-yl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 1-(1-Boc-piperidin-4-yl)pyrazole-4-boronic Acid Pinacol Ester
PubChem CID 45480279
IUPAC Name tert-butyl 4-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrazol-1-yl]piperidine-1-carboxylate
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CN(N=C2)C3CCN(CC3)C(=O)OC(C)(C)C

Specifications

Melting Point 116°C
Color White
Quantity 1 g
Formula Weight 377.29
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material tert-Butyl 4-[4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)pyrazol-1-yl]piperidine-1-carboxylate
TSCA No
Recommended Storage Refrigerator