missing translation for 'onlineSavingsMsg'
Learn More
Learn More
tert-Butyl 4-(2-formylphenyl)piperazine-1-carboxylate, 97%, Thermo Scientific™
Chemical Identifiers
| CAS | 174855-57-3 |
|---|---|
| Molecular Formula | C16H22N2O3 |
| Molecular Weight (g/mol) | 290.363 |
| MDL Number | MFCD05864664 |
| InChI Key | FGJACYJASSSXNJ-UHFFFAOYSA-N |
| Synonym | 1-boc-4-2-formylphenyl piperazine, tert-butyl 4-2-formylphenyl piperazine-1-carboxylate, 4-2-formylphenyl piperazine-1-carboxylic acid tert-butyl ester, tert-butyl 4-2-formylphenyl piperazinecarboxylate, 4-2-formyl-phenyl-piperazine-1-carboxylic acid tert-butyl ester, acmc-20a0zo, n-boc-4-2-formylphenyl-piperazine, 1-n-boc-4-2-formylphenyl piperazine, 1-boc-4-2-formyl-phenyl-piperazine |
| PubChem CID | 6490990 |
| IUPAC Name | tert-butyl 4-(2-formylphenyl)piperazine-1-carboxylate |
| SMILES | CC(C)(C)OC(=O)N1CCN(CC1)C2=CC=CC=C2C=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|
CC30704DA
|
N/A | 1g | Amber Glass Bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
| 174855-57-3 | |
| 290.363 | |
| FGJACYJASSSXNJ-UHFFFAOYSA-N | |
| 6490990 | |
| CC(C)(C)OC(=O)N1CCN(CC1)C2=CC=CC=C2C=O |
| C16H22N2O3 | |
| MFCD05864664 | |
| 1-boc-4-2-formylphenyl piperazine, tert-butyl 4-2-formylphenyl piperazine-1-carboxylate, 4-2-formylphenyl piperazine-1-carboxylic acid tert-butyl ester, tert-butyl 4-2-formylphenyl piperazinecarboxylate, 4-2-formyl-phenyl-piperazine-1-carboxylic acid tert-butyl ester, acmc-20a0zo, n-boc-4-2-formylphenyl-piperazine, 1-n-boc-4-2-formylphenyl piperazine, 1-boc-4-2-formyl-phenyl-piperazine | |
| tert-butyl 4-(2-formylphenyl)piperazine-1-carboxylate |
Specifications
| 174855-57-3 | |
| 100.0 | |
| C16H22N2O3 | |
| 1-boc-4-2-formylphenyl piperazine, tert-butyl 4-2-formylphenyl piperazine-1-carboxylate, 4-2-formylphenyl piperazine-1-carboxylic acid tert-butyl ester, tert-butyl 4-2-formylphenyl piperazinecarboxylate, 4-2-formyl-phenyl-piperazine-1-carboxylic acid tert-butyl ester, acmc-20a0zo, n-boc-4-2-formylphenyl-piperazine, 1-n-boc-4-2-formylphenyl piperazine, 1-boc-4-2-formyl-phenyl-piperazine | |
| CC(C)(C)OC(=O)N1CCN(CC1)C2=CC=CC=C2C=O | |
| 290.363 | |
| 290.36 |
| 96.5 | |
| 97% | |
| MFCD05864664 | |
| FGJACYJASSSXNJ-UHFFFAOYSA-N | |
| tert-butyl 4-(2-formylphenyl)piperazine-1-carboxylate | |
| 6490990 | |
| Tert-butyl 4-(2-formylphenyl)piperazine-1-carboxylate |