missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl 4-(2-chloroethyl)piperazine-1-carboxylate, 97%

Catalog No. AC438240050
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC438240050 5 g Glass bottle
AC438240010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC438240050 Supplier Thermo Scientific Chemicals Supplier No. 438240050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 208167-83-3 | C11H21ClN2O2 | 248.76 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 208167-83-3
Molecular Formula C11H21ClN2O2
Molecular Weight (g/mol) 248.76
InChI Key MYOWELLYEZMECA-UHFFFAOYSA-N
Synonym tert-butyl 4-2-chloroethyl piperazine-1-carboxylate, tert-butyl 4-2-chloroethyl tetrahydro-1 2h-pyrazinecarboxylate, 1-boc-4-2-chloroethyl piperazine, tert-butyl 4-2-chloroethyl piperazinecarboxylate, 1-piperazinecarboxylic acid, 4-2-chloroethyl-, 1,1-dimethylethyl ester, 4-2-chloroethyl piperazine, n1-boc protected, 4-2-chloroethyl piperazine,n1-boc protected, tertbutylchloroethyltetrahydropyrazinecarboxylate, tert-butyl 4-2-chloroethyl tetrahydro-1 2h-pyraz
PubChem CID 22106269
IUPAC Name tert-butyl 4-(2-chloroethyl)piperazine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCN(CC1)CCCl

Specifications

Melting Point 62.9°C to 64.7°C
Color White to Brown
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 5 g
Formula Weight 248.76
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material tert-Butyl 4-(2-chloroethyl)piperazine-1-carboxylate
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Harmful if inhaled.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: If breathing is difficult,remove to fresh air and keep at rest in a position comfortable for breathing.

RUO – Research Use Only