missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl (3S)-3-aminobutanoate, 95%, Thermo Scientific Chemicals

$183.53 - $183.53

Chemical Identifiers

CAS 161105-54-0
MDL Number MFCD00798294
InChI Key BFFNZGWJTHWUMY-LURJTMIESA-N
PubChem CID 7021083
IUPAC Name tert-butyl (3S)-3-aminobutanoate
SMILES CC(CC(=O)OC(C)(C)C)N
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC337941000
SDS View Documents
Thermo Scientific Chemicals
337941000
100 mg
Each for $183.53
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

161105-54-0
BFFNZGWJTHWUMY-LURJTMIESA-N
tert-butyl (3S)-3-aminobutanoate
MFCD00798294
7021083
CC(CC(=O)OC(C)(C)C)N

Specifications

161105-54-0
Authentic
C8H17NO2
1.4230 to 1.427
+ 24
BFFNZGWJTHWUMY-LURJTMIESA-N
tert-butyl (3S)-3-aminobutanoate
159.23
159.23
Liquid
Colorless to Yellow
94% min. (GC)
Glass Bottle
MFCD00798294
tert-butyl 3s-3-aminobutanoate, s-tert-butyl 3-aminobutanoate, butanoic acid, 3-amino-, 1,1-dimethylethyl ester, 3s, tert-butyl-3s-3-aminobutanoate, tert-butyl s-3-aminobutyrate, 1,1-dimethylethyl 35-3-aminobutanoate, s-3-aminobutyric acid tert-butyl ester, 1,1-dimethylethyl 3s-3-aminobutanoate, butanoic acid,3-amino-, 1,1-dimethylethyl ester, 3s
CC(CC(=O)OC(C)(C)C)N
100 mg
7021083
95%
tert-Butyl (3S)-3-aminobutanoate, 95%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS
Documents

Documents

RUO – Research Use Only