missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl (3S)-3-amino-4-phenylbutanoate, 97%

CAS: 120686-17-1 | C14H22NO2 | 236.33 g/mol

Supplier:  Thermo Scientific Chemicals 344465000

 View more versions of this product

Catalog No. AC344465000


Only null left

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

tert-Butyl (3S)-3-amino-4-phenylbutanoate
120686-17-1
Authentic
C14H22NO2
MFCD00798308
s-tert-butyl 3-amino-4-phenylbutanoate, tert-butyl 3s-3-amino-4-phenylbutanoate, t-butyl 3s-3-amino-4-phenylbutanoate, 1,1-dimethylethyl 3s-3-amino-4-phenylbutanoate, benzenebutanoic acid, b-amino-, 1,1-dimethylethyl ester, bs, t-butyl 3r-3-amino-4-phenylbutanoate, s-tert-butyl3-amino-4-phenylbutanoate, tert-butyl 3s-3-amino-4-phenyl-butanoate, s-3-amino-4-phenyl-butyric acid tert-butyl ester, s-3-amino-4-phenylbutanoic acid tert-butyl ester
CC(C)(C)OC(=O)C[C@H]([NH3+])CC1=CC=CC=C1
500 mg
1501868
97%
97%
White or Yellow
96% min. (GC)
Glass Bottle
2.3
ZIJHIHDFXCNFAA-GFCCVEGCSA-O
tert-butyl (3S)-3-amino-4-phenylbutanoate
236.33
235.3
Low Melting Solid

Chemical Identifiers

120686-17-1
236.33
ZIJHIHDFXCNFAA-GFCCVEGCSA-O
tert-butyl (3S)-3-amino-4-phenylbutanoate
C14H22NO2
MFCD00798308
1501868
CC(C)(C)OC(=O)C[C@H]([NH3+])CC1=CC=CC=C1
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only