missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl (3S)-3-amino-4-phenylbutanoate, 97%

Catalog No. p-81220
Change view
Click to view available options
Quantity:
100 mg
500 mg
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC344461000 100 mg Glass Bottle
AC344465000 500 mg Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC344461000 Supplier Thermo Scientific Chemicals Supplier No. 344461000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 120686-17-1 | C14H22NO2 | 236.33 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 120686-17-1
Molecular Formula C14H22NO2
Molecular Weight (g/mol) 236.33
MDL Number MFCD00798308
InChI Key ZIJHIHDFXCNFAA-GFCCVEGCSA-O
PubChem CID 1501868
IUPAC Name tert-butyl (3S)-3-amino-4-phenylbutanoate
SMILES CC(C)(C)OC(=O)C[C@H]([NH3+])CC1=CC=CC=C1

Specifications

Color White or Yellow
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Quantity 100 mg
Specific Rotation 2.3
Synonym s-tert-butyl 3-amino-4-phenylbutanoate, tert-butyl 3s-3-amino-4-phenylbutanoate, t-butyl 3s-3-amino-4-phenylbutanoate, 1,1-dimethylethyl 3s-3-amino-4-phenylbutanoate, benzenebutanoic acid, b-amino-, 1,1-dimethylethyl ester, bs, t-butyl 3r-3-amino-4-phenylbutanoate, s-tert-butyl3-amino-4-phenylbutanoate, tert-butyl 3s-3-amino-4-phenyl-butanoate, s-3-amino-4-phenyl-butyric acid tert-butyl ester, s-3-amino-4-phenylbutanoic acid tert-butyl ester
Formula Weight 235.3
Percent Purity 97%
Physical Form Low Melting Solid
Chemical Name or Material tert-Butyl (3S)-3-amino-4-phenylbutanoate, 97%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only