missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl (3S)-3-amino-3-phenylpropanoate, 97%

Catalog No. p-81202
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC337892500 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC337892500 Supplier Thermo Scientific Chemicals Supplier No. 337892500
Only null left

CAS: 120686-18-2 | C13H20NO2 | 222.31 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 120686-18-2
Molecular Formula C13H20NO2
Molecular Weight (g/mol) 222.31
MDL Number MFCD00798310
InChI Key TYYCBAISLMKLMT-NSHDSACASA-O
PubChem CID 7021093
SMILES CC(C)(C)OC(=O)C[C@H]([NH3+])C1=CC=CC=C1

Specifications

Color Colorless to Brown
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass Bottle
Refractive Index 1.4960 to 1.498
Specific Rotation -22
Synonym s-tert-butyl 3-amino-3-phenylpropanoate, tert-butyl 3s-3-amino-3-phenylpropanoate, benzenepropanoic acid, b-amino-, 1,1-dimethylethyl ester, bs, s-tert-butyl3-amino-3-phenylpropanoate, t-butyl 3s-3-amino-3-phenylpropanoate, s-alpha-2-oxo-2-tert-butoxyethyl benzyl amine, tert-butyl-3s-3-amino-3-phenylpropanoate, benzenepropanoic acid,b-amino-,1,1-dimethylethyl ester, bs
Specific Rotation Condition - 22.00 (20.00°C c=1,CHCl3)
IUPAC Name tert-butyl (3S)-3-amino-3-phenylpropanoate
Quantity 250 mg
Formula Weight 221.3
Percent Purity 97%
Physical Form Slightly Viscous Liquid
Chemical Name or Material tert-Butyl (3S)-3-amino-3-phenylpropanoate, 97%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only