missing translation for 'onlineSavingsMsg'
Learn More
Learn More
tert-Butyl 3-Oxopiperazine-1-carboxylate 98.0+%, TCI America™
$57.79 - $156.59
Chemical Identifiers
| CAS | 76003-29-7 |
|---|---|
| Molecular Formula | C9H16N2O3 |
| Molecular Weight (g/mol) | 200.238 |
| MDL Number | MFCD02181069 |
| InChI Key | FCMLWBBLOASUSO-UHFFFAOYSA-N |
| Synonym | 1-boc-3-oxopiperazine, 4-n-boc-2-oxo-piperazine, 4-boc-piperazinone, n-boc-3-oxopiperazine, 3-oxo-piperazine-1-carboxylic acid tert-butyl ester, tert-butyl 3-oxo-1-piperazinecarboxylate, 4-n-boc-piperazin-2-one, n-boc-piperazin-3-one, 1-piperazinecarboxylic acid, 3-oxo-, 1,1-dimethylethyl ester, 1-tert-butoxycarbonyl-3-oxopiperazine |
| PubChem CID | 3157178 |
| IUPAC Name | tert-butyl 3-oxopiperazine-1-carboxylate |
| SMILES | CC(C)(C)OC(=O)N1CCNC(=O)C1 |
Chemical Identifiers
| 76003-29-7 | |
| 200.238 | |
| FCMLWBBLOASUSO-UHFFFAOYSA-N | |
| 3157178 | |
| CC(C)(C)OC(=O)N1CCNC(=O)C1 |
| C9H16N2O3 | |
| MFCD02181069 | |
| 1-boc-3-oxopiperazine, 4-n-boc-2-oxo-piperazine, 4-boc-piperazinone, n-boc-3-oxopiperazine, 3-oxo-piperazine-1-carboxylic acid tert-butyl ester, tert-butyl 3-oxo-1-piperazinecarboxylate, 4-n-boc-piperazin-2-one, n-boc-piperazin-3-one, 1-piperazinecarboxylic acid, 3-oxo-, 1,1-dimethylethyl ester, 1-tert-butoxycarbonyl-3-oxopiperazine | |
| tert-butyl 3-oxopiperazine-1-carboxylate |
Specifications
| 76003-29-7 | |
| White-Yellow | |
| MFCD02181069 | |
| 1-boc-3-oxopiperazine, 4-n-boc-2-oxo-piperazine, 4-boc-piperazinone, n-boc-3-oxopiperazine, 3-oxo-piperazine-1-carboxylic acid tert-butyl ester, tert-butyl 3-oxo-1-piperazinecarboxylate, 4-n-boc-piperazin-2-one, n-boc-piperazin-3-one, 1-piperazinecarboxylic acid, 3-oxo-, 1,1-dimethylethyl ester, 1-tert-butoxycarbonyl-3-oxopiperazine | |
| CC(C)(C)OC(=O)N1CCNC(=O)C1 | |
| 200.238 | |
| 200.24 | |
| Crystalline Powder |
| 159°C | |
| C9H16N2O3 | |
| 1 g | |
| FCMLWBBLOASUSO-UHFFFAOYSA-N | |
| tert-butyl 3-oxopiperazine-1-carboxylate | |
| 3157178 | |
| ≥98.0% (GC) | |
| tert-Butyl 3-Oxopiperazine-1-carboxylate |
Safety and Handling
TSCA : No