missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl 3-Oxopiperazine-1-carboxylate 98.0+%, TCI America™

Catalog No. B38045G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B38045G 5 g
B38041G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B38045G Supplier TCI America Supplier No. B38045G

Chemical Identifiers

CAS 76003-29-7
Molecular Formula C9H16N2O3
Molecular Weight (g/mol) 200.238
MDL Number MFCD02181069
InChI Key FCMLWBBLOASUSO-UHFFFAOYSA-N
Synonym 1-boc-3-oxopiperazine, 4-n-boc-2-oxo-piperazine, 4-boc-piperazinone, n-boc-3-oxopiperazine, 3-oxo-piperazine-1-carboxylic acid tert-butyl ester, tert-butyl 3-oxo-1-piperazinecarboxylate, 4-n-boc-piperazin-2-one, n-boc-piperazin-3-one, 1-piperazinecarboxylic acid, 3-oxo-, 1,1-dimethylethyl ester, 1-tert-butoxycarbonyl-3-oxopiperazine
PubChem CID 3157178
IUPAC Name tert-butyl 3-oxopiperazine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCNC(=O)C1

Specifications

Melting Point 159°C
Color White-Yellow
Quantity 5 g
Formula Weight 200.24
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material tert-Butyl 3-Oxopiperazine-1-carboxylate