missing translation for 'onlineSavingsMsg'
Learn More

tert-butyl 3-endo-hydroxy-8-azabicyclo[3.2.1]octane-8-carboxylate, Thermo Scientific™

Catalog No. CC74315DA
Change view
Click to view available options
Quantity:
1g
5g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC74315DA 1g Amber Glass Bottle
CC74315DE 5g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC74315DA Supplier Thermo Scientific Chemicals Supplier No. CC74315DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 143557-91-9
Molecular Formula C12H21NO3
Molecular Weight (g/mol) 227.304
MDL Number MFCD09966165
InChI Key SEGZJJSZYOEABC-IDKOKCKLSA-N
PubChem CID 58414652
IUPAC Name tert-butyl (5S)-3-hydroxy-8-azabicyclo[3.2.1]octane-8-carboxylate
SMILES CC(C)(C)OC(=O)N1C2CCC1CC(C2)O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1g
Formula Weight 227.3
Percent Purity 97%
Chemical Name or Material Tert-butyl 3-endo-hydroxy-8-azabicyclo[3.2.1]octane-8-carboxylate
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Harmful if swallowed.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.