missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl 3-Carbamoylpyrrolidine-1-carboxylate 98.0+%, TCI America™

Catalog No. B52485G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B52485G 5 g
B52481G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B52485G Supplier TCI America Supplier No. B52485G
Only null left

Chemical Identifiers

CAS 122684-34-8
Molecular Formula C10H18N2O3
Molecular Weight (g/mol) 214.265
MDL Number MFCD04116210
InChI Key NHDGOVOBEZPXMY-UHFFFAOYSA-N
Synonym 3-aminocarbonyl-1-boc-pyrrolidine, 1-boc-3-carbamoylpyrrolidine, 1-boc-pyrrolidine-3-carboxamide, 3-carbamoyl-pyrrolidine-1-carboxylic acid tert-butyl ester, 1-boc-pyrrolidine-3-carboxyamide, tert-butyl 3-aminocarbonyl pyrrolidine-1-carboxylate, acmc-20a2cc, ksc496a4t, 1-boc-3-pyrrolidinecarboxamide, 1-n-boc-pyrrolidine-3-carboxyamide
PubChem CID 4571096
IUPAC Name tert-butyl 3-carbamoylpyrrolidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCC(C1)C(=O)N

Specifications

Melting Point 119°C
Color White-Yellow
Quantity 5 g
Formula Weight 214.27
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material tert-Butyl 3-Carbamoylpyrrolidine-1-carboxylate