missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl 2-Bromoisobutyrate 98.0+%, TCI America™

Catalog No. B350025G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B350025G 25 g
B3500500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B350025G Supplier TCI America Supplier No. B350025G

Chemical Identifiers

CAS 23877-12-5
Molecular Formula C8H15BrO2
Molecular Weight (g/mol) 223.11
MDL Number MFCD00051908
InChI Key IGVNJALYNQVQIT-UHFFFAOYSA-N
Synonym tert-butyl 2-bromoisobutyrate, t-butyl 2-bromo isobutyrate, 2-bromoisobutyric acid tert-butylester, tert-butyl 2-bromo-2-methylpropionate, tert-butyl alpha-bromoisobutyrate, t-butyl 2-bromoisobutylate, t-butyl 2-bromoisobutyrate, 2-bromoisobutyric acid tert-butyl ester, propanoic acid, 2-bromo-2-methyl-, 1,1-dimethylethyl ester, t-butyl2-bromoisobutyrate
PubChem CID 90290
IUPAC Name tert-butyl 2-bromo-2-methylpropanoate
SMILES CC(C)(C)OC(=O)C(C)(C)Br

Specifications

Color Yellow
Boiling Point 37°C
Quantity 25 g
Formula Weight 223.11
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material tert-Butyl 2-Bromoisobutyrate
EINECSNumber (2)-3333
TSCA No