missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl 2-bromobutyrate, 98%

Catalog No. p-7046284
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1379906 5 g
AAL1379914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1379906 Supplier Thermo Scientific Chemicals Supplier No. L1379906
Only null left

CAS: 24457-21-4 | C8H15BrO2 | 223.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 24457-21-4
Molecular Formula C8H15BrO2
Molecular Weight (g/mol) 223.11
MDL Number MFCD00209634
InChI Key RIUJWUWLGXBICR-UHFFFAOYSA-N
Synonym tert-butyl 2-bromobutyrate, tert-butyl2-bromobutanoate, t-butyl 2-bromobutyrate, 2-bromobutyric acid tert-butyl ester, 2-bromobutanoic acid tert-butyl ester, butanoic acid, 2-bromo-, 1,1-dimethylethyl ester, tertbutyl 2-bromobutyrate, tert-butyl-2-bromobutyrat, acmc-1cb0d, tert-butyl-2-bromobutyrate
PubChem CID 13124118
IUPAC Name tert-butyl 2-bromobutanoate
SMILES CCC(C(=O)OC(C)(C)C)Br

Specifications

Density 1.2
Boiling Point 72°C to 76°C (12 mmHg)
Refractive Index 1.441
Quantity 5 g
UN Number UN1760
Beilstein 2040298
Formula Weight 223.12
Percent Purity 98%
Chemical Name or Material tert-Butyl 2-bromobutyrate
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only