missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl 2-bromo-6,7-dihydro[1,3]thiazolo[5,4-c]pyridine-5(4H)-carboxylate, 97%, Thermo Scientific Chemicals

Catalog No. p-7077424
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC453590050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC453590050 Supplier Thermo Scientific Chemicals Supplier No. 453590050

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 365996-06-1
Molecular Formula C11H15BrN2O2S
Molecular Weight (g/mol) 319.22
InChI Key RXXKNZKFRAFFOH-UHFFFAOYSA-N
Synonym tert-butyl 2-bromo-6,7-dihydrothiazolo 5,4-c pyridine-5 4h-carboxylate, 5-boc-2-bromo-6,7-dihydro-4h-thiazolo 5,4-c pyridine, tert-butyl 2-bromo-6,7-dihydro 1,3 thiazolo 5,4-c pyridine-5 4h-carboxylate, tert-butyl 2-bromo-4h,5h,6h,7h-1,3 thiazolo 5,4-c pyridine-5-carboxylate, 2-bromo-6,7-dihydro-4h-thiazolo 5,4-c pyridine-5-carboxylic acid tert-butyl ester, 2-bromo-4,5,6,7-tetrahydro 1,3 thiazolo 5,4-c pyridine, n-boc protected, tert-butyl 2-bromo-4h,6h,7h-1,3 thiazolo 5,4-c pyridine-5-carboxylate, pubchem15507, tert-butyl 2-bromo-6,7-dihydrothiazolo 5,4-c pyrid, 5-n-boc-2-bromo-4,5,6,7-tetrahydrothiazolo 5,4-c pyridine
PubChem CID 11162951
IUPAC Name tert-butyl 2-bromo-6,7-dihydro-4H-[1,3]thiazolo[5,4-c]pyridine-5-carboxylate
SMILES CC(C)(C)OC(=O)N1CCC2=C(C1)SC(=N2)Br

Specifications

Melting Point 94.0°C to 97.0°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Quantity 5 g
Formula Weight 319.22
Percent Purity 97%
Chemical Name or Material tert-Butyl 2-bromo-6,7-dihydro[1,3]thiazolo[5,4-c]pyridine-5(4H)-carboxylate
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.
Precautionary Statement GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Call a POISON CENTER or doctor/physician if you feel unwell.

RUO – Research Use Only