missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl 2-aminoisobutyrate hydrochloride, 98%

Catalog No. AAH6381703
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6381703 1 g
AAH6381706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6381703 Supplier Thermo Scientific Chemicals Supplier No. H6381703
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 84758-81-6 | C8H17NO2 | 159.23 g/mol

tert-Butyl 2-aminoisobutyrate hydrochloride is used as intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
tert-Butyl 2-aminoisobutyrate hydrochloride is used as intermediates.

Solubility
Soluble in water or 1% acetic acid.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, heat.

Chemical Identifiers

CAS 84758-81-6
Molecular Formula C8H17NO2
Molecular Weight (g/mol) 159.23
MDL Number MFCD00237698
InChI Key LSVYCJILORYVCD-UHFFFAOYSA-N
Synonym alpha-aminoisobutyric acid t-butyl ester, h-aib-otbu.hcl, tert-butyl 2-methylalaninate, 2-aminoisobutyric acid t-butyl ester, alanine, 2-methyl-, 1,1-dimethylethyl ester, 2-amino-2-methylpropionic acid tert-butyl ester, h-aib-otbu inverted exclamation mark currencyhcl, t-butyl 2-methylalaninate, 2-methylalanine t-butyl ester, h-alpha-me-ala-otbu . hcl
PubChem CID 7019940
IUPAC Name tert-butyl 2-amino-2-methylpropanoate
SMILES CC(C)(C)OC(=O)C(C)(C)N

Specifications

Color White
Quantity 1 g
Solubility Information Soluble in water or 1% acetic acid.
Formula Weight 195.69
Percent Purity 98%
Physical Form Powder
Chemical Name or Material tert-Butyl 2-aminoisobutyrate hydrochloride
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold

RUO – Research Use Only