missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl 2,2,2-Trichloroacetimidate 95.0+%, TCI America™

Catalog No. B14965ML
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B14965ML 5 mL
B149625ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B14965ML Supplier TCI America Supplier No. B14965ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 98946-18-0
Molecular Formula C6H10Cl3NO
Molecular Weight (g/mol) 218.50
MDL Number MFCD00077410
InChI Key CQXDYHPBXDZWBA-UHFFFAOYSA-N
Synonym tert-butyl 2,2,2-trichloroacetimidate, tert-butyl trichloroacetimidate, tert-butyl-2,2,2-trichloroacetimidate, tert-butyl 2,2,2-trichloroethanecarboximidate, 2,2,2-trichloroacetimidic acid tert-butyl ester, t-butyl trichloroacetimidate, t-butyl 2,2,2-trichloroacetimidate, tert-butyl2,2,2-trichloroacetimidate, ethanimidic acid, 2,2,2-trichloro-, 1,1-dimethylethyl ester, 1-tert-butoxy-2,2,2-trichloroethanimine
PubChem CID 2734700
IUPAC Name tert-butyl 2,2,2-trichloroethanimidate
SMILES CC(C)(C)OC(=N)C(Cl)(Cl)Cl

Specifications

Color Yellow
Boiling Point 69°C
Quantity 5 mL
UN Number 1993
Formula Weight 218.50
Percent Purity ≥95.0% (GC)
Grade Reagent
Physical Form Liquid
Chemical Name or Material tert-Butyl 2,2,2-Trichloroacetimidate
TSCA No
Recommended Storage Refrigerator