missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl 1-piperazinecarboxylate, 97%

Catalog No. AC367280250
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC367280250 25 g Glass bottle
AC367280050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC367280250 Supplier Thermo Scientific Chemicals Supplier No. 367280250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 57260-71-6 | C9H19N2O2 | 187.26 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 57260-71-6
Molecular Formula C9H19N2O2
Molecular Weight (g/mol) 187.26
MDL Number MFCD00075265
InChI Key CWXPZXBSDSIRCS-UHFFFAOYSA-O
Synonym 1-boc-piperazine, n-boc-piperazine, 1-n-boc-piperazine, tert-butyl 1-piperazinecarboxylate, 1-tert-butoxycarbonyl piperazine, n-t-butoxycarbonylpiperazine, boc-piperazine, n-tert-butoxycarbonylpiperazine, piperazine-1-carboxylic acid tert-butyl ester, 1-piperazinecarboxylic acid, 1,1-dimethylethyl ester
PubChem CID 143452
IUPAC Name tert-butyl piperazine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CC[NH2+]CC1

Specifications

Melting Point 43°C to 47°C
Color White to Yellow
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 25 g
Beilstein 23, V,1, 55
Formula Weight 186.25
Percent Purity 97%
Physical Form Crystalline Low Melting Solid
Chemical Name or Material tert-Butyl 1-piperazinecarboxylate
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minu

RUO – Research Use Only