Learn More
(tert-Butoxycarbonylmethylene)triphenylphosphorane, 98%
CAS: 35000-38-5 | C24H25O2P | 376.44 g/mol
$146.00 - $531.36
Chemical Identifiers
| CAS | 35000-38-5 |
|---|---|
| Molecular Formula | C24H25O2P |
| Molecular Weight (g/mol) | 376.44 |
| MDL Number | MFCD00075545 |
| InChI Key | ZWZUFQPXYVYAFO-UHFFFAOYSA-N |
| Synonym | tert-butyl 2-triphenylphosphoranylidene acetate, tert-butoxycarbonylmethylene triphenylphosphorane, tert-butyl triphenylphosphoranylidene acetate, acetic acid, triphenylphosphoranylidene-, 1,1-dimethylethyl ester, t-butoxycarbonylmethylene triphenylphosphorane, tert-butyl 2-triphenyl-??-phosphanylidene acetate, triphenylthoranylidene acetic acid 1,1-dimethylethyl ester, pubchem14371, tert-butyl 2-triphenyl, tert-butyl 2-triphenylylidene acetate |
| PubChem CID | 256127 |
| IUPAC Name | tert-butyl 2-(triphenyl-$l^{5}-phosphanylidene)acetate |
| SMILES | CC(C)(C)OC(=O)C=P(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAL1514406
|
Thermo Scientific Chemicals
L1514406 |
5 g |
Each for $146.00
|
|
|||||
|
AAL1514414
|
Thermo Scientific Chemicals
L1514414 |
25 g |
Each for $531.36
|
|
|||||
Description
(tert-Butoxycarbonylmethylene)triphenylphosphorane is a chemical used in the synthesis of aldose reductase inhibitors, a podophyllotoxin derivative and tautomycin, a podophyllotoxin derivative and tautomycin. It Is also used in medicinal chemistry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications(tert-Butoxycarbonylmethylene)triphenylphosphorane is a chemical used in the synthesis of aldose reductase inhibitors, a podophyllotoxin derivative and tautomycin, a podophyllotoxin derivative and tautomycin. It Is also used in medicinal chemistry.
Solubility
Sparingly soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature is 2 - 8°C. Incompatible with strong oxidizing agents.
Chemical Identifiers
| 35000-38-5 | |
| 376.44 | |
| ZWZUFQPXYVYAFO-UHFFFAOYSA-N | |
| 256127 | |
| CC(C)(C)OC(=O)C=P(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1 |
| C24H25O2P | |
| MFCD00075545 | |
| tert-butyl 2-triphenylphosphoranylidene acetate, tert-butoxycarbonylmethylene triphenylphosphorane, tert-butyl triphenylphosphoranylidene acetate, acetic acid, triphenylphosphoranylidene-, 1,1-dimethylethyl ester, t-butoxycarbonylmethylene triphenylphosphorane, tert-butyl 2-triphenyl-??-phosphanylidene acetate, triphenylthoranylidene acetic acid 1,1-dimethylethyl ester, pubchem14371, tert-butyl 2-triphenyl, tert-butyl 2-triphenylylidene acetate | |
| tert-butyl 2-(triphenyl-$l^{5}-phosphanylidene)acetate |
Specifications
| 35000-38-5 | |
| C24H25O2P | |
| MFCD00075545 | |
| UN2811 | |
| tert-butyl 2-triphenylphosphoranylidene acetate, tert-butoxycarbonylmethylene triphenylphosphorane, tert-butyl triphenylphosphoranylidene acetate, acetic acid, triphenylphosphoranylidene-, 1,1-dimethylethyl ester, t-butoxycarbonylmethylene triphenylphosphorane, tert-butyl 2-triphenyl-??-phosphanylidene acetate, triphenylthoranylidene acetic acid 1,1-dimethylethyl ester, pubchem14371, tert-butyl 2-triphenyl, tert-butyl 2-triphenylylidene acetate | |
| ZWZUFQPXYVYAFO-UHFFFAOYSA-N | |
| tert-butyl 2-(triphenyl-$l^{5}-phosphanylidene)acetate | |
| 256127 | |
| 98% |
| 146°C to 150°C | |
| (CH3)3CO2CCH=P(C6H5)3 | |
| 5 g | |
| 2759875 | |
| Sparingly soluble in water. | |
| CC(C)(C)OC(=O)C=P(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1 | |
| 376.44 | |
| 376.44 | |
| (tert-Butoxycarbonylmethylene)triphenylphosphorane |
Safety and Handling
GHS H Statement
H301-H373-H319-H317
Toxic if swallowed.
May cause damage to organs through prolonged or repeated exposure.
Causes serious eye irritation.
May cause an allergic skin reaction.
P260-P264b-P270-P272-P280-P301+P310-P302+P352-P305+P351+P338-P314-P330-P333+P313-P363-P501c
H301-H317-H319-H373
DOTInformation : Hazard Class: 6.1; Packaging Group: II
EINECSNumber : 412-880-7
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only