missing translation for 'onlineSavingsMsg'
Learn More

(tert-Butoxycarbonylmethylene)triphenylphosphorane, 98%

Catalog No. AAL1514406
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1514406 5 g
AAL1514414 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1514406 Supplier Thermo Scientific Chemicals Supplier No. L1514406
Only null left

CAS: 35000-38-5 | C24H25O2P | 376.44 g/mol

(tert-Butoxycarbonylmethylene)triphenylphosphorane is a chemical used in the synthesis of aldose reductase inhibitors, a podophyllotoxin derivative and tautomycin, a podophyllotoxin derivative and tautomycin. It Is also used in medicinal chemistry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(tert-Butoxycarbonylmethylene)triphenylphosphorane is a chemical used in the synthesis of aldose reductase inhibitors, a podophyllotoxin derivative and tautomycin, a podophyllotoxin derivative and tautomycin. It Is also used in medicinal chemistry.

Solubility
Sparingly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature is 2 - 8°C. Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 35000-38-5
Molecular Formula C24H25O2P
Molecular Weight (g/mol) 376.44
MDL Number MFCD00075545
InChI Key ZWZUFQPXYVYAFO-UHFFFAOYSA-N
Synonym tert-butyl 2-triphenylphosphoranylidene acetate, tert-butoxycarbonylmethylene triphenylphosphorane, tert-butyl triphenylphosphoranylidene acetate, acetic acid, triphenylphosphoranylidene-, 1,1-dimethylethyl ester, t-butoxycarbonylmethylene triphenylphosphorane, tert-butyl 2-triphenyl-??-phosphanylidene acetate, triphenylthoranylidene acetic acid 1,1-dimethylethyl ester, pubchem14371, tert-butyl 2-triphenyl, tert-butyl 2-triphenylylidene acetate
PubChem CID 256127
IUPAC Name tert-butyl 2-(triphenyl-$l^{5}-phosphanylidene)acetate
SMILES CC(C)(C)OC(=O)C=P(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 146°C to 150°C
Linear Formula (CH3)3CO2CCH=P(C6H5)3
Quantity 5 g
UN Number UN2811
Beilstein 2759875
Solubility Information Sparingly soluble in water.
Formula Weight 376.44
Percent Purity 98%
Chemical Name or Material (tert-Butoxycarbonylmethylene)triphenylphosphorane
Hazard Category H301-H317-H319-H373
Hazard Statement GHS H Statement
H301-H373-H319-H317
Toxic if swallowed.
May cause damage to organs through prolonged or repeated exposure.
Causes serious eye irritation.
May cause an allergic skin reaction.
Precautionary Statement P260-P264b-P270-P272-P280-P301+P310-P302+P352-P305+P351+P338-P314-P330-P333+P313-P363-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: II
EINECSNumber 412-880-7
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only