missing translation for 'onlineSavingsMsg'
Learn More

(tert-Butoxycarbonylmethyl)triphenylphosphonium Bromide 98.0+%, TCI America™

Catalog No. B392825G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B392825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B392825G Supplier TCI America Supplier No. B392825G
Only null left

Chemical Identifiers

CAS 59159-39-6
Molecular Formula C24H26BrO2P
Molecular Weight (g/mol) 457.35
MDL Number MFCD00075527
InChI Key ZGLFRTJDWWKIAK-UHFFFAOYSA-M
Synonym tert-butoxycarbonylmethyl triphenylphosphonium bromide, tert-butoxycarbonylmethyl triphenylphosphanium bromide, 2-tert-butoxy-2-oxoethyl triphenylphosphonium bromide, phosphonium, 2-1,1-dimethylethoxy-2-oxoethyl triphenyl-, bromide, 2-tert-butoxy-2-oxoethyl triphenylphosphanium bromide, acmc-20apek, ksc609k1l, carbo-t-butoxymethyl triphenylphosphonium bromide, tert-butoxy carbonyl methyl triphenyl phosphoniu
PubChem CID 2733853
IUPAC Name [2-(tert-butoxy)-2-oxoethyl]triphenylphosphanium bromide
SMILES [Br-].CC(C)(C)OC(=O)C[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 176°C
Color White
Quantity 25 g
Formula Weight 457.35
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material (tert-Butoxycarbonylmethyl)triphenylphosphonium Bromide