missing translation for 'onlineSavingsMsg'
Learn More

[(tert-Butoxycarbonyl)aminooxy]acetic Acid 98.0+%, TCI America™

Catalog No. p-7122885
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B33381G 1 g
B33385G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B33381G Supplier TCI America Supplier No. B33381G

Chemical Identifiers

CAS 42989-85-5
Molecular Formula C7H13NO5
Molecular Weight (g/mol) 191.183
MDL Number MFCD01632027
InChI Key QBXODCKYUZNZCY-UHFFFAOYSA-N
Synonym boc-aminooxy acetic acid, 2-tert-butoxycarbonyl amino oxy acetic acid, boc-aoa-oh, boc-aminooxyacetic acid, boc-aoac-oh, n-boc-carboxymethoxy amine, boc-aminooxy aceticacid, 2-tert-butoxy carbonyl amino oxy acetic acid, 2-2-methylpropan-2-yl oxycarbonylamino oxyacetic acid, tert-butoxycarbonyl aminooxy acetic acid
PubChem CID 2755974
IUPAC Name 2-[(2-methylpropan-2-yl)oxycarbonylamino]oxyacetic acid
SMILES CC(C)(C)OC(=O)NOCC(=O)O

Specifications

Melting Point 115°C
Color White-Yellow
Quantity 1 g
Formula Weight 191.18
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material [(tert-Butoxycarbonyl)aminooxy]acetic Acid