missing translation for 'onlineSavingsMsg'
Learn More

tert-Butanol, CHROMASOLV™, for HPLC, ≥99.5%, Honeywell Riedel-de Haën™

Catalog No. p-7111801
Change view
Click to view available options
Quantity:
2.5 L
4 x 2.5 L
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
6001641 2.5 L Glass Bottle
6001642 4 x 2.5 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 6001641 Supplier Honeywell Chemicals Supplier No. 3082502.5L
Only null left

CHROMASOLV™, for HPLC,≥99.5%

  • For HPLC and organic synthesis
  • High-purity, multipurpose solvents
  • High lot-to-lot consistency

Chemical Identifiers

CAS 75-65-0
Molecular Formula C4H10O
Molecular Weight (g/mol) 74.123
MDL Number MFCD00004464
InChI Key DKGAVHZHDRPRBM-UHFFFAOYSA-N
Synonym tert-butanol, tert-butyl alcohol, 2-methyl-2-propanol, t-butanol, t-butyl hydroxide, 1,1-dimethylethanol, trimethylcarbinol, trimethyl methanol, 2-propanol, 2-methyl, t-butyl alcohol
PubChem CID 6386
ChEBI CHEBI:45895
IUPAC Name 2-methylpropan-2-ol
SMILES CC(C)(C)O

Specifications

Melting Point 23°C to 26°C (literature)
Absorbance max. 0.01 at 350 nm, max. 0.01 at 300 nm, max. 0.20 at 250 nm, max. 0.50 at 230 nm, max. 1.00 at 215 nm
CAS Max % 100.0000%
Density 0.775 g/mL at 25 °C(lit.)
Boiling Point 81°C to 83°C
Quantity 2.5 L
Flash Point 11°C
Packaging Glass Bottle
Linear Formula (CH3)3COH
UN Number UN1120
Beilstein 906698
Formula Weight 74.12
Percent Purity >99.5%
Grade HPLC
Chemical Name or Material tert-Butanol, CHROMASOLV™, for HPLC, ≥99.5%
Show More Show Less
ShelfLife 720 days from date of manufacture
DOTInformation Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: Butanols
EINECSNumber 200-889-7
Recommended Storage Room Temp