missing translation for 'onlineSavingsMsg'
Learn More

tert-Butanol, 99.5%, extra pure

Catalog No. p-3735808
Change view
Click to view available options
Quantity:
1 L
2.5 L
10 L
Packaging:
Plastic Bottle
Plastic Drum
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC107710010 1 L Plastic Bottle
AC107710025 2.5 L Plastic Bottle
AC107710100 10 L Plastic Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC107710010 Supplier Thermo Scientific Chemicals Supplier No. 107710010
Only null left

CAS: 75-65-0 | C4H10O | 74.123 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 75-65-0
Molecular Formula C4H10O
Molecular Weight (g/mol) 74.123
MDL Number MFCD00004464
InChI Key DKGAVHZHDRPRBM-UHFFFAOYSA-N
Synonym tert-butanol, tert-butyl alcohol, 2-methyl-2-propanol, t-butanol, t-butyl hydroxide, 1,1-dimethylethanol, trimethylcarbinol, trimethyl methanol, 2-propanol, 2-methyl, t-butyl alcohol
PubChem CID 6386
ChEBI CHEBI:45895
IUPAC Name 2-methylpropan-2-ol
SMILES CC(C)(C)O

Specifications

Melting Point 25.0°C to 25.5°C
Color Colorless
Density 0.7800g/mL
Boiling Point 83.0°C
Quantity 1 L
Flash Point 11°C
Infrared Spectrum Authentic
Packaging Plastic Bottle
Physical Form Liquid After Melting
Refractive Index 1.3860 to 1.3880
Assay Percent Range 99.5%
Residue after Evaporation 0.1% max.
Specific Gravity 0.78
Linear Formula (CH3)3COH
Viscosity 3.35 mPa.s (30°C)
Beilstein 01, 379
Water 0.03% max. (K.F.)
Merck Index 15, 1544
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol, ether, acetone
Formula Weight 74.12
Percent Purity 99.5%
Grade Extra Pure
Chemical Name or Material tert-Butanol
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if inhaled.
Highly flammable liquid and vapor.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only