missing translation for 'onlineSavingsMsg'
Learn More

tert-Amylamine 98.0+%, TCI America™

Catalog No. A100225ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A100225ML 25 mL
A1002500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A100225ML Supplier TCI America Supplier No. A100225ML

Chemical Identifiers

CAS 594-39-8
Molecular Formula C5H13N
Molecular Weight (g/mol) 87.166
MDL Number MFCD00008056
InChI Key GELMWIVBBPAMIO-UHFFFAOYSA-N
Synonym tert-amylamine, tert-pentylamine, 2-butanamine, 2-methyl, t-amylamine, 1,1-dimethylpropylamine, 2-methyl-2-butylamine, 2-amino-2-methylbutane, 2-methyl-butan-2-amine, 1,1-dimethyl-1-propylamine, t-pentylamine
PubChem CID 68986
ChEBI CHEBI:84236
IUPAC Name 2-methylbutan-2-amine
SMILES CCC(C)(C)N

Specifications

Color Yellow
Boiling Point 77°C
Quantity 25 mL
UN Number 1106
Formula Weight 87.17
Percent Purity ≥98.0% (GC,T)
Grade Reagent
Physical Form Liquid
Chemical Name or Material tert-Amylamine
EINECSNumber (2)-0133
TSCA Yes