missing translation for 'onlineSavingsMsg'
Learn More

tert-Amyl Alcohol 98.0+%, TCI America™

Catalog No. p-7134401
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P005925ML 25 mL
P0059500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P005925ML Supplier TCI America Supplier No. P005925ML
Only null left

Chemical Identifiers

CAS 75-85-4
Molecular Formula C5H12O
Molecular Weight (g/mol) 88.15
MDL Number MFCD00004478
InChI Key MSXVEPNJUHWQHW-UHFFFAOYSA-N
Synonym 2-methyl-2-butanol, tert-amyl alcohol, amylene hydrate, tert-pentyl alcohol, tert-pentanol, t-amyl alcohol, 2-butanol, 2-methyl, dimethylethylcarbinol, ethyldimethylcarbinol, tert-isoamyl alcohol
PubChem CID 6405
IUPAC Name 2-methylbutan-2-ol
SMILES CCC(C)(C)O

Specifications

Melting Point -8°C
Color Colorless
Boiling Point 102°C
Quantity 25 mL
UN Number 1105
Formula Weight 88.15
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material tert-Amyl Alcohol
EINECSNumber (2)-0217
RTECSNumber SC0175000
TSCA Yes