missing translation for 'onlineSavingsMsg'
Learn More

Fisher BioReagents™ Terrific Broth (Powder/Molecular Genetics)

Catalog No. BP2468500
Change view
Click to view available options
Quantity:
10,000 g
500 g
2000 g
Packaging Type:
Poly Bottle
Poly Pail
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging Type
BP2468500 500 g Poly Bottle
BP24682 2000 g Poly Bottle
BP246810 10,000 g Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. BP2468500 Supplier Fisher BioReagents™ Supplier No. BP2468 500
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Enriched medium used with glycerol in cultivating recombinant strains of Escherichia coli.

Formulation Usage: 50.8g/L

Specifications

Content And Storage 4°C
Form Powder
Grade Molecular genetics
Media Type Terrific Broth
Product Type Microbiology Media
For Use With (Application) Cultivation of recombinant strains of Escherichia coli
Packaging Type Poly Bottle
Quantity 500 g