missing translation for 'onlineSavingsMsg'
Learn More

Fisher BioReagents™ Terrific Broth, Modified (Granulated)

Catalog No. p-4245455
Change view
Click to view available options
Quantity:
500 g
2 kg
Packaging Type:
Poly Bottle
Catalog No. Quantity Packaging Type
BP9728500 500 g Poly Bottle
BP97282 2 kg Poly Bottle
2 options

Catalog No. BP9728500

Supplier: Fisher BioReagents™ BP9728 500

Only null left

Used as a microbial growth medium for the cultivation of E. coli strains for the preparation of plasmid DNA and recombinant proteins.

  • Directions for Media Preparation: Add 47.6g to 1L of purified water, such as BP2820 (Fisher BioReagents Microbial Cell Culture Grade Water). Then, add 4 mL of glycerol to the mixture. Heat and frequently agitate to completely dissolve. Before use, sterilize by autoclaving for 15 minutes.
  • Greater peptone and yeast concentration allow for higher plasmid yield
  • Format: Granulated powder

Specifications

Form Granulated Powder
Grade Molecular genetics
Media Type Terrific Broth
Product Type Microbiology Media
Formulation Casein Peptone 12g/L, Yeast extract 24g/L, Potassium Phosphate, Monobasic 2.2g/L, Potassium Phosphate, Dibasic 9.4g/L
For Use With (Application) For maintenance and propagation of Escherichia coli in molecular biology procedures
Packaging Type Poly Bottle
Quantity 500 g