missing translation for 'onlineSavingsMsg'
Learn More

Research Products Corporation Terrific Broth, Modified, Granulated, 5 Kilograms

Catalog No. 50489058
Encompass

Enriched media for maintenance and propagation of E. Coli and plasmid growth. Produces higher cell densities resulting in greater yields of DNA or recombinant problem. Glycerol is used as a carbon source with this formulation instead of glucose as it will not ferment to acetic acid.

Ingredients:

Casein Digest Peptone: 12g/l
Yeast Extract: 24g/l
Dipotassium phosphate: 9.5g/l
Monopotassium phosphate: 2.2g/

Catalog No. 50489058 Supplier Research Products Corporation Supplier No. RPI T151005000.0

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Enriched media for maintenance and propagation of E. Coli and plasmid growth. Produces higher cell densities resulting in greater yields of DNA or recombinant problem. Glycerol is used as a carbon source with this formulation instead of glucose as it will not ferment to acetic acid.

Ingredients:

Casein Digest Peptone: 12g/l
Yeast Extract: 24g/l
Dipotassium phosphate: 9.5g/l
Monopotassium phosphate: 2.2g/