missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Terpinyl Acetate 85.0+%, TCI America™
$31.68 - $132.35
Chemical Identifiers
| CAS | 80-26-2 |
|---|---|
| Molecular Formula | C12H20O2 |
| Molecular Weight (g/mol) | 196.29 |
| MDL Number | MFCD00037155 |
| InChI Key | IGODOXYLBBXFDW-UHFFFAOYNA-N |
| Synonym | alpha-terpinyl acetate, terpineol acetate, terpineol, acetate, p-menth-1-en-8-yl acetate, p-menth-1-en-8-ol, acetate, alpha-terpineol, acetate, .alpha.-terpinyl acetate, alpha-terpineol acetate, +/--alpha-terpinyl acetate, 3-cyclohexene-1-methanol, .alpha.,.alpha.,4-trimethyl-, acetate |
| PubChem CID | 111037 |
| ChEBI | CHEBI:32320 |
| IUPAC Name | 2-(4-methylcyclohex-3-en-1-yl)propan-2-yl acetate |
| SMILES | CC(=O)OC(C)(C)C1CCC(C)=CC1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
T002325ML
|
TCI America
T002325ML |
25 mL |
Each for $31.68
|
|
|||||
|
T0023500ML
|
TCI America
T0023500ML |
500 mL |
Each for $132.35
|
|
|||||
Chemical Identifiers
| 80-26-2 | |
| 196.29 | |
| IGODOXYLBBXFDW-UHFFFAOYNA-N | |
| 111037 | |
| 2-(4-methylcyclohex-3-en-1-yl)propan-2-yl acetate |
| C12H20O2 | |
| MFCD00037155 | |
| alpha-terpinyl acetate, terpineol acetate, terpineol, acetate, p-menth-1-en-8-yl acetate, p-menth-1-en-8-ol, acetate, alpha-terpineol, acetate, .alpha.-terpinyl acetate, alpha-terpineol acetate, +/--alpha-terpinyl acetate, 3-cyclohexene-1-methanol, .alpha.,.alpha.,4-trimethyl-, acetate | |
| CHEBI:32320 | |
| CC(=O)OC(C)(C)C1CCC(C)=CC1 |
Specifications
| 80-26-2 | |
| 94°C | |
| MFCD00037155 | |
| alpha-terpinyl acetate, terpineol acetate, terpineol, acetate, p-menth-1-en-8-yl acetate, p-menth-1-en-8-ol, acetate, alpha-terpineol, acetate, .alpha.-terpinyl acetate, alpha-terpineol acetate, +/--alpha-terpinyl acetate, 3-cyclohexene-1-methanol, .alpha.,.alpha.,4-trimethyl-, acetate | |
| CC(=O)OC(C)(C)C1CCC(C)=CC1 | |
| 196.29 | |
| CHEBI:32320 | |
| ≥85.0% (GC) | |
| Terpinyl Acetate |
| Colorless | |
| C12H20O2 | |
| 25 mL | |
| IGODOXYLBBXFDW-UHFFFAOYNA-N | |
| 2-(4-methylcyclohex-3-en-1-yl)propan-2-yl acetate | |
| 111037 | |
| 196.29 | |
| Liquid |
Safety and Handling
EINECSNumber : (3)-2358
RTECSNumber : OT0200000
TSCA : Yes