missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Terpinolene 90.0+%, TCI America™
$50.33 - $137.94
Chemical Identifiers
| CAS | 586-62-9 |
|---|---|
| Molecular Formula | C10H16 |
| Molecular Weight (g/mol) | 136.24 |
| MDL Number | MFCD00049191 |
| InChI Key | MOYAFQVGZZPNRA-UHFFFAOYSA-N |
| Synonym | terpinolene, isoterpinene, terpinolen, alpha-terpinolene, 1,4 8-p-menthadiene, p-mentha-1,4 8-diene, 4-isopropylidene-1-methylcyclohexene, tereben, p-menth-1,4 8-diene, cyclohexene, 1-methyl-4-1-methylethylidene |
| PubChem CID | 11463 |
| ChEBI | CHEBI:9457 |
| IUPAC Name | 1-methyl-4-(propan-2-ylidene)cyclohex-1-ene |
| SMILES | CC(C)=C1CCC(C)=CC1 |
Chemical Identifiers
| 586-62-9 | |
| 136.24 | |
| MOYAFQVGZZPNRA-UHFFFAOYSA-N | |
| 11463 | |
| 1-methyl-4-(propan-2-ylidene)cyclohex-1-ene |
| C10H16 | |
| MFCD00049191 | |
| terpinolene, isoterpinene, terpinolen, alpha-terpinolene, 1,4 8-p-menthadiene, p-mentha-1,4 8-diene, 4-isopropylidene-1-methylcyclohexene, tereben, p-menth-1,4 8-diene, cyclohexene, 1-methyl-4-1-methylethylidene | |
| CHEBI:9457 | |
| CC(C)=C1CCC(C)=CC1 |
Specifications
| 586-62-9 | |
| 185°C | |
| MFCD00049191 | |
| 2541 | |
| MOYAFQVGZZPNRA-UHFFFAOYSA-N | |
| 1-methyl-4-(propan-2-ylidene)cyclohex-1-ene | |
| 11463 | |
| 136.24 | |
| Liquid |
| Yellow | |
| C10H16 | |
| 25 mL | |
| terpinolene, isoterpinene, terpinolen, alpha-terpinolene, 1,4 8-p-menthadiene, p-mentha-1,4 8-diene, 4-isopropylidene-1-methylcyclohexene, tereben, p-menth-1,4 8-diene, cyclohexene, 1-methyl-4-1-methylethylidene | |
| CC(C)=C1CCC(C)=CC1 | |
| 136.24 | |
| CHEBI:9457 | |
| ≥90.0% (GC) | |
| Terpinolene |
Safety and Handling
EINECSNumber : (3)-2226&(3)-2228
RTECSNumber : WZ6870000
TSCA : Yes