missing translation for 'onlineSavingsMsg'
Learn More

Terpinolene 90.0+%, TCI America™

Catalog No. T0817500ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T0817500ML 500 mL
T081725ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T0817500ML Supplier TCI America Supplier No. T0817500ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 586-62-9
Molecular Formula C10H16
Molecular Weight (g/mol) 136.24
MDL Number MFCD00049191
InChI Key MOYAFQVGZZPNRA-UHFFFAOYSA-N
Synonym terpinolene, isoterpinene, terpinolen, alpha-terpinolene, 1,4 8-p-menthadiene, p-mentha-1,4 8-diene, 4-isopropylidene-1-methylcyclohexene, tereben, p-menth-1,4 8-diene, cyclohexene, 1-methyl-4-1-methylethylidene
PubChem CID 11463
ChEBI CHEBI:9457
IUPAC Name 1-methyl-4-(propan-2-ylidene)cyclohex-1-ene
SMILES CC(C)=C1CCC(C)=CC1

Specifications

Color Yellow
Boiling Point 185°C
Quantity 500 mL
UN Number 2541
Formula Weight 136.24
Percent Purity ≥90.0% (GC)
Physical Form Liquid
Chemical Name or Material Terpinolene
EINECSNumber (3)-2226&(3)-2228
RTECSNumber WZ6870000
TSCA Yes