missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Terpinolene 90.0+%, TCI America™
Specifications
Specifications
| Color | Yellow |
| Boiling Point | 185°C |
| Quantity | 25 mL |
| UN Number | 2541 |
| Formula Weight | 136.24 |
| Percent Purity | ≥90.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | Terpinolene |
Safety and Handling
| EINECSNumber | (3)-2226&(3)-2228 |
| RTECSNumber | WZ6870000 |
| TSCA | Yes |