missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Tergitol™ 15-S-9

Catalog No. AC464250010
Change view
Click to view available options
Quantity:
250 mL
1 L
2.5 L
10 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC464250010 1 L
AC464252500 250 mL
AC464250025 2.5 L
AC464250100 10 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC464250010 Supplier Thermo Scientific Chemicals Supplier No. 464250010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 68131-40-8 | [C2H4O]n C13H27O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 68131-40-8
Molecular Formula [C2H4O]n C13H27O
MDL Number MFCD00132411
SMILES CCCCCCCC(CCCCC)OCCO-*

Specifications

Density 1.0100 g/mL
Quantity 1 L
Flash Point 193°C
Specific Gravity 1.01
Solubility Information Solubility aq. soln.: soluble.
Chemical Name or Material Tergitol* 15-S-9
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement Harmful if swallowed.

Harmful if inhaled.

Causes skin irritation.

Causes serious eye damage.

May cause respiratory irritation.

Precautionary Statement GHS P Statement IF SWALLOWED: rinse mouth.

Do NOT induce vomiting.

IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.

IF ON SKIN: Wash with plenty of soap and water.

IF IN EYES: Rinse cautiously with water for several minutes.

Remove contact lenses, if present and easy to do.

Continue rinsing.

Immediately call a POISON CENTER or doctor/physician.

Wear protective gloves/protective clothing/eye protection/face protection.

RUO – Research Use Only