missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Terfenadine

Catalog No. AAJ61936MC
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ61936MC 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ61936MC Supplier Thermo Scientific Chemicals Supplier No. J61936MC

CAS: 50679-08-8 | C32H41NO2

Terfenadine has been used to study the role of histamine in itch related to proteinase-activated receptors (PARs) in mice. Terfenadine has also been used to block histamine receptor type 1 to study the pathogenesis of 2,4-dinitrobenzene sulfonic acid (DNBS)-induced ulcerative colitis in rats.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 50679-08-8
Molecular Formula C32H41NO2

Specifications

Quantity 100 mg
Description White to Off-white
Content And Storage 2°C to 8°C
For Use With (Application) An antihistamine
Formula Weight 471.67g/mol
Melting Point 145°C to 152°C
Product Type Signal Transduction Reagent
Form Powder

RUO – Research Use Only