missing translation for 'onlineSavingsMsg'
Learn More

Terephthaloyl Chloride 99.0+%, TCI America™

Catalog No. T0017500G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T0017500G 500 g
T001725G 25 g
T0017100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. T0017500G Supplier TCI America Supplier No. T0017500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 100-20-9
Molecular Formula C8H4Cl2O2
Molecular Weight (g/mol) 203.02
MDL Number MFCD00000693
InChI Key LXEJRKJRKIFVNY-UHFFFAOYSA-N
Synonym terephthaloyl chloride, terephthaloyl dichloride, 1,4-benzenedicarbonyl dichloride, terephthalic acid dichloride, terephthalic dichloride, p-phthaloyl chloride, terephthalic acid chloride, p-phthalyl dichloride, p-phthaloyl dichloride, terephthalyl dichloride
PubChem CID 7488
IUPAC Name benzene-1,4-dicarbonyl dichloride
SMILES ClC(=O)C1=CC=C(C=C1)C(Cl)=O

Specifications

Melting Point 83°C
Color White
Boiling Point 266°C
Quantity 500 g
UN Number 2923
Formula Weight 203.02
Percent Purity ≥99.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Terephthaloyl Chloride
EINECSNumber (3)-1378
RTECSNumber WZ1797000
TSCA Yes