missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Terephthalonitrile 98.0+%, TCI America™
$80.85 - $658.41
Chemical Identifiers
| CAS | 623-26-7 |
|---|---|
| Molecular Formula | C8H4N2 |
| Molecular Weight (g/mol) | 128.134 |
| MDL Number | MFCD00001810 |
| InChI Key | BHXFKXOIODIUJO-UHFFFAOYSA-N |
| Synonym | terephthalonitrile, 1,4-dicyanobenzene, 1,4-benzenedicarbonitrile, p-phthalodinitrile, 4-cyanobenzonitrile, p-dicyanobenzene, p-benzenedinitrile, p-pdn, terephthalodinitrile, 1,4-benzodinitrile |
| PubChem CID | 12172 |
| IUPAC Name | benzene-1,4-dicarbonitrile |
| SMILES | C1=CC(=CC=C1C#N)C#N |
Chemical Identifiers
| 623-26-7 | |
| 128.134 | |
| BHXFKXOIODIUJO-UHFFFAOYSA-N | |
| 12172 | |
| C1=CC(=CC=C1C#N)C#N |
| C8H4N2 | |
| MFCD00001810 | |
| terephthalonitrile, 1,4-dicyanobenzene, 1,4-benzenedicarbonitrile, p-phthalodinitrile, 4-cyanobenzonitrile, p-dicyanobenzene, p-benzenedinitrile, p-pdn, terephthalodinitrile, 1,4-benzodinitrile | |
| benzene-1,4-dicarbonitrile |
Specifications
| 623-26-7 | |
| White | |
| MFCD00001810 | |
| terephthalonitrile, 1,4-dicyanobenzene, 1,4-benzenedicarbonitrile, p-phthalodinitrile, 4-cyanobenzonitrile, p-dicyanobenzene, p-benzenedinitrile, p-pdn, terephthalodinitrile, 1,4-benzodinitrile | |
| C1=CC(=CC=C1C#N)C#N | |
| 128.134 | |
| 128.13 | |
| Crystalline Powder |
| 228°C | |
| C8H4N2 | |
| 25 g | |
| BHXFKXOIODIUJO-UHFFFAOYSA-N | |
| benzene-1,4-dicarbonitrile | |
| 12172 | |
| ≥98.0% (GC) | |
| Terephthalonitrile |
Safety and Handling
EINECSNumber : (3)-1799
RTECSNumber : CZ1925000
TSCA : Yes