missing translation for 'onlineSavingsMsg'
Learn More

Terephthalonitrile 98.0+%, TCI America™

Catalog No. T0016500G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T0016500G 500 g
T001625G 25 g
T0016100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. T0016500G Supplier TCI America Supplier No. T0016500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 623-26-7
Molecular Formula C8H4N2
Molecular Weight (g/mol) 128.134
MDL Number MFCD00001810
InChI Key BHXFKXOIODIUJO-UHFFFAOYSA-N
Synonym terephthalonitrile, 1,4-dicyanobenzene, 1,4-benzenedicarbonitrile, p-phthalodinitrile, 4-cyanobenzonitrile, p-dicyanobenzene, p-benzenedinitrile, p-pdn, terephthalodinitrile, 1,4-benzodinitrile
PubChem CID 12172
IUPAC Name benzene-1,4-dicarbonitrile
SMILES C1=CC(=CC=C1C#N)C#N

Specifications

Melting Point 228°C
Color White
Quantity 500 g
Formula Weight 128.13
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Terephthalonitrile
EINECSNumber (3)-1799
RTECSNumber CZ1925000
TSCA Yes