missing translation for 'onlineSavingsMsg'
Learn More

Terephthalamide 98.0+%, TCI America™

Catalog No. T001325G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T001325G 25 g
T0013500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T001325G Supplier TCI America Supplier No. T001325G

Chemical Identifiers

CAS 3010-82-0
Molecular Formula C8H8N2O2
Molecular Weight (g/mol) 164.16
MDL Number MFCD00025482
InChI Key MHSKRLJMQQNJNC-UHFFFAOYSA-N
PubChem CID 76381
ChEBI CHEBI:38802
IUPAC Name benzene-1,4-dicarboxamide
SMILES NC(=O)C1=CC=C(C=C1)C(N)=O

Specifications

Color White
Quantity 25 g
Formula Weight 164.16
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material Terephthalamide
RTECSNumber CZ2202000
TSCA Yes