missing translation for 'onlineSavingsMsg'
Learn More

Terephthalamic Acid 95.0+%, TCI America™

Catalog No. T17685G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T17685G 5 g
T176825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T17685G Supplier TCI America Supplier No. T17685G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 6051-43-0
Molecular Formula C8H7NO3
Molecular Weight (g/mol) 165.15
MDL Number MFCD02258929
InChI Key JMHSCWJIDIKGNZ-UHFFFAOYSA-N
Synonym Terephthalamidic Acid
PubChem CID 506057
ChEBI CHEBI:50738
IUPAC Name 4-carbamoylbenzoic acid
SMILES NC(=O)C1=CC=C(C=C1)C(O)=O

Specifications

Melting Point 248°C
Color White
Quantity 5 g
Formula Weight 165.15
Percent Purity ≥95.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Terephthalamic Acid