missing translation for 'onlineSavingsMsg'
Learn More

Terephthalaldehydic Acid 98.0+%, TCI America™

Catalog No. p-7136026
Change view
Click to view available options
Quantity:
100 g
25 g
500 g
Catalog No. Quantity
T001125G 25 g
T0011100G 100 g
T0011500G 500 g
3 options

Catalog No. T001125G

Supplier: TCI America T001125G

Only null left

Chemical Identifiers

CAS 619-66-9
Molecular Formula C8H6O3
Molecular Weight (g/mol) 150.133
MDL Number MFCD00006951
InChI Key GOUHYARYYWKXHS-UHFFFAOYSA-N
Synonym 4-carboxybenzaldehyde, benzoic acid, 4-formyl, terephthalaldehydic acid, p-formylbenzoic acid, p-carboxybenzaldehyde, terephthaldehydic acid, 4-carboxybenzaladehyde, 4-formyl-benzoic acid, 4-formylbenzoicacid, unii-ues4qrk36e
PubChem CID 12088
IUPAC Name 4-formylbenzoic acid
SMILES C1=CC(=CC=C1C=O)C(=O)O

Specifications

Melting Point 256°C
Color White-Yellow
Quantity 25 g
Formula Weight 150.13
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Terephthalaldehydic Acid
EINECSNumber (3)-1670
RTECSNumber WZ0440000
TSCA Yes