missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Terbutaline Impurity C, British Pharmacopoeia (BP) Reference Standard, MilliporeSigma™ Supelco™
Specifications
Specifications
| CAS | 139508-58-0 |
| Molecular Formula | C12H17NO3 |
| Linear Formula | C12H17NO3 |
| Quantity | 15 mg |
| Synonym | 1-(3,5-Dihydroxyphenyl)-2-[(1,1-dimethylethyl)amino]ethanone; Terbutalone |
| Formula Weight | 223.27 |
| Grade | British Pharmacopoeia (BP) Reference Standard |