missing translation for 'onlineSavingsMsg'
Learn More

Terbium(III) trifluoromethanesulfonate, 98%

Catalog No. p-7051030
Change view
Click to view available options
Quantity:
2 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4031204 2 g
AA4031209 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4031204 Supplier Thermo Scientific Chemicals Supplier No. 04031204

CAS: 148980-31-8 | C3F9O9S3Tb | 606.115 g/mol

It is used as a reagent & chemical intermediates. It is a raw material used in synthesis of chemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 148980-31-8
Molecular Formula C3F9O9S3Tb
Molecular Weight (g/mol) 606.115
MDL Number MFCD00192461
InChI Key JLYAVYSVLLIWEK-UHFFFAOYSA-K
Synonym terbium iii trifluoromethanesulfonate, terbium tritriflate, terbium iii triflate, terbium tris trifluoromethanesulfonate, trifluoromethanesulfonic acid terbium iii salt, terbium trifluoromethanesulfonate, terbium triflate, tb otf 3, acmc-1byi6
PubChem CID 11786785
IUPAC Name terbium(3+);trifluoromethanesulfonate
SMILES C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].[Tb+3]

Specifications

Physical Form Powder
Quantity 2 g
Solubility Information Insoluble in water.
Formula Weight 606.12
Percent Purity 98%
Chemical Name or Material Terbium(III) trifluoromethanesulfonate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only