missing translation for 'onlineSavingsMsg'
Learn More

Terbium(III) chloride hexahydrate, REacton™, 99.9% (REO)

Catalog No. p-4408185
Change view
Click to view available options
Quantity:
10 g
50 g
Catalog No. Quantity
AA1120909 10 g
AA1120918 50 g
2 options

Catalog No. AA1120909

Supplier: Thermo Scientific Chemicals 01120909

Only null left

CAS: 13798-24-8 | Cl3H12O6Tb | 373.37 g/mol

Terbium(III) chloride hexahydrate is a water soluble source used for the preparation of metallic terbium, which is used in electronic devices and an additive for special alloy namely Terfenol-D. It is used in the preparation of complexes, which are used in the spectrofluorimetric determination of various chemicals in pharmaceutical formulations. Further, it finds use in the preparation of terbium nanotubes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 13798-24-8
Molecular Formula Cl3H12O6Tb
Molecular Weight (g/mol) 373.37
MDL Number MFCD00149870
InChI Key ULJUVCOAZNLCJZ-UHFFFAOYSA-K
IUPAC Name terbium(3+) hexahydrate trichloride
SMILES O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Tb+3]

Specifications

Physical Form Crystalline Aggregates
Quantity 10 g
Assay Percent Range 99.9% (REO)
Solubility Information Very soluble in water.
Formula Weight 373.38 (265.29 Anhydrous)
Sensitivity Hygroscopic
Chemical Name or Material Terbium(III) chloride hexahydrate
EINECSNumber 233-132-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only