missing translation for 'onlineSavingsMsg'
Learn More

Terbium(III) chloride hexahydrate, 99.9%, (trace metal basis)

CAS: 13798-24-8 | Cl3H12O6Tb | 373.37 g/mol

$168.70 - $168.70

Chemical Identifiers

CAS 13798-24-8
Molecular Formula Cl3H12O6Tb
Molecular Weight (g/mol) 373.37
MDL Number MFCD00149870
InChI Key ULJUVCOAZNLCJZ-UHFFFAOYSA-K
IUPAC Name terbium(3+) hexahydrate trichloride
SMILES O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Tb+3]
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC199610050
SDS View Documents
Thermo Scientific Chemicals
199610050
5 g Glass bottle
Each for $168.70
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for ultra trace analysis
Specifications

Chemical Identifiers

13798-24-8
373.37
ULJUVCOAZNLCJZ-UHFFFAOYSA-K
O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Tb+3]
Cl3H12O6Tb
MFCD00149870
terbium(3+) hexahydrate trichloride

Specifications

White
Crystalline Powder, Crystals and/or Chunks
99.90%
TbCl3·6H2O
14, 9157
ULJUVCOAZNLCJZ-UHFFFAOYSA-K
terbium(3+) hexahydrate trichloride
373.38
Trace Metal Basis
Terbium(III) chloride hexahydrate
13798-24-8
5 g
Cl3H12O6Tb
MFCD00149870
Solubility in water: soluble. Other solubilities: readily soluble in alcohol
O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Tb+3]
373.37
99.9%
Glass bottle
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only